Learn More
TARGETMOL CHEMICALS INC DESOXYRHAPONTICIN 10MG
SDP
Catalog No. 502578910
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Desoxyrhaponticin (Deoxyrhaponticin) is an agent that is potentially effective in controlling postprandial hyperglycemia in diabetes the in vivo antidiabetic action of this compound can be explained in part at least by inhibition of glucose transport in the small intestine and inhibition of glucose reabsorption in the kidney. 2. Desoxyrhaponticin and rhaponticin could be considered as promising fatty acid synthase(FAS) inhibitors they could inhibit intracellular FAS activity and downregulate FAS expression in human breast cancer MCF-7 cells. purity: 99%
Catalog No. 50-257-8910 Supplier TARGETMOL CHEMICALS INC Supplier No. TJS038710MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Desoxyrhaponticin (Deoxyrhaponticin) is an agent that is potentially effective in controlling postprandial hyperglycemia in diabetes the in vivo antidiabetic action of this compound can be explained in part at least by inhibition of glucose transport in the small intestine and inhibition of glucose reabsorption in the kidney. 2. Desoxyrhaponticin and rhaponticin could be considered as promising fatty acid synthase(FAS) inhibitors they could inhibit intracellular FAS activity and downregulate FAS expression in human breast cancer MCF-7 cells. purity: 99%