Learn More
Medchemexpress LLC HY-N2486 5mg Medchemexpress, Desoxyrhaponticin CAS:30197-14-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN24865MG
Encompass_Preferred
Medchemexpress, HY-N2486 5mg Desoxyrhaponticin CAS:30197-14-9 Desoxyrhaponticin is a stilbene glycoside from the Tibetan nutritional food Rheum tanguticum Maxim. Desoxyrhaponticin is a Fatty acid synthase (FAS) inhibitor, and has apoptotic effect on human cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0397