Learn More
Grainger Dessert Dish 3 3/4 in Dia 5 oz PK24
Catalog No. 502750931
Encompass_Preferred
The Grainger 14D105 is a Westward brand 1/4-inch drive click-type torque wrench. It has a torque range of 10 to 50 inch pounds. The wrench has a reversible ratchet head that allows operation in tight spaces. It features a knurled grip handle and a clearly marked, high contrast dual scale. The wrench has an accuracy of ±4%. It is made from chrome vanadium steel for durability. The wrench comes pre-calibrated from the factory and is ready to use out of the package. It weighs 0.60 lbs. This click-type torque wrench provides reliable torque control for tightening fasteners to exact specifications.
Catalog No. 50-275-0931 Supplier Grainger Supplier No. 14D105
The Grainger 14D105 is a Westward brand 1/4-inch drive click-type torque wrench. It has a torque range of 10 to 50 inch pounds. The wrench has a reversible ratchet head that allows operation in tight spaces. It features a knurled grip handle and a clearly marked, high contrast dual scale. The wrench has an accuracy of ±4%. It is made from chrome vanadium steel for durability. The wrench comes pre-calibrated from the factory and is ready to use out of the package. It weighs 0.60 lbs. This click-type torque wrench provides reliable torque control for tightening fasteners to exact specifications.