Learn More
Electron Microscopy Sciences Silica Gel Bulk Desiccant, 2 Lbs
SDP
Catalog No. NC1513274
Encompass_Preferred
Changes color when it reaches max absorption capacity.
Catalog No. NC1513274 Supplier Electron Microscopy Sciences Supplier No. 7120507
Changes color when it reaches max absorption capacity.