Learn More
Parker Hannifan Parker Hannifin MKH2PEM-D Desiccant Cartridge Kit, Moisture Removal, Compatible with H2PEM Generators, Unit Count: 1
SDP
Catalog No. NC9810614
Encompass
The Parker Hannifin desiccant cartridge kit ensures ultra-high purity hydrogen by effectively removing moisture from gas streams, making it essential for analytical hydrogen generators. With compatible models including the Parker H2PEM series, it helps maintain gas purity levels between 99.9999% to 99.99999+%. The cartridge supports applications in precise gas quality monitoring and regularly integrates into 6-month and 24-month service kits for optimal performance.
Catalog No. NC9810614 Supplier Parker Hannifan Supplier No. MKH2PEMD
The Parker Hannifin desiccant cartridge kit ensures ultra-high purity hydrogen by effectively removing moisture from gas streams, making it essential for analytical hydrogen generators. With compatible models including the Parker H2PEM series, it helps maintain gas purity levels between 99.9999% to 99.99999+%. The cartridge supports applications in precise gas quality monitoring and regularly integrates into 6-month and 24-month service kits for optimal performance.