Learn More
Parker Hannifan DESSICANT CARTRIDGE WITH BEADS
SDP
Catalog No. NC9569022
Encompass
Dessicant Cartridge, For Hydrogen Generators, Fuel Grade Purity: 99.9995%(around )
Catalog No. NC9569022 Supplier Parker Hannifan Supplier No. 1647727
Dessicant Cartridge, For Hydrogen Generators, Fuel Grade Purity: 99.9995%(around )