Learn More
Perkin Elmer US LLC Dessicant Dryer Cartridge
Catalog No. 502016587
Encompass_Preferred
Dessicant Dryer Cartridge Technical Data Cartridge: -100°F Dew Point Duration (at Recommended Flow): 420 minutes Volume Flow: 2100 Ft3 Max Surface Velocity: 71 Ft/Min Recommended Contact Time: 10 sec Recommended Flow Capacity at 100 psig: 0.1 Ft3/Min to 5 Ft3/Min
Catalog No. 50-201-6587 Supplier Perkin Elmer US LLC Supplier No. 09220018
Dessicant Dryer Cartridge Technical Data Cartridge: -100°F Dew Point Duration (at Recommended Flow): 420 minutes Volume Flow: 2100 Ft3 Max Surface Velocity: 71 Ft/Min Recommended Contact Time: 10 sec Recommended Flow Capacity at 100 psig: 0.1 Ft3/Min to 5 Ft3/Min