Learn More
Chemglass Life Sciences Desiccator, 223.5mm I.D., 5.8L, Large Knob Top, 190mm Plate Dia, Complete
SDP
Catalog No. NC1448469
Encompass_Preferred
This 5.8L Pyrex desiccator is designed for general analytical work. The large, sturdy knob makes the cover easy to handle. Approximate bowl volume is 5,800mL. For replacement cover see CG-8266-R200CO. For lower part only see CG-8266-R200LO. Please Note: Plate is not supplied and must be purchased seperately.WARNING: Do Not Heat Or Subject To Pressure. Not For Vacuum Applications.
Catalog No. NC1448469 Supplier Chemglass Life Sciences Supplier No. CG8266R200
This 5.8L Pyrex desiccator is designed for general analytical work. The large, sturdy knob makes the cover easy to handle. Approximate bowl volume is 5,800mL. For replacement cover see CG-8266-R200CO. For lower part only see CG-8266-R200LO. Please Note: Plate is not supplied and must be purchased seperately.WARNING: Do Not Heat Or Subject To Pressure. Not For Vacuum Applications.