Learn More
Hach Company Desiccator, Non-vacuum
Supplier:  Hach Company 1428500
Encompass
Round polycarbonate cover and polypropylene base. Non-Vacuum. Chamber is impervious to alkali and acidic conditions. Recessed rim prevents grease (not included) from contacting bench surfaces when removed and eliminates need for a separate gasket. Dimensions (height x diam): 245 mm x 330 mm (9.7" x 13"). Holds 454 g (1 lb) of dessicant. Ceramic dessicator plate (sold separately) is inert, non-stick, and corrosion resistant. Quadrants crafted into plate for easy tracking of material locations. Plate diameter: 230 mm (9").
Catalog No. 50-199-4984