Learn More
Hach Company Hach Desiccator Cabinet, Non-Vacuum - Stackable Polystyrene (21 x 18 x 13 cm)
Catalog No. NC3781094 Shop All Hach Company Products
Encompass
The desiccator cabinet, non-vacuum provides an airtight, stackable storage solution for moisture-sensitive samples and components. Constructed of transparent polystyrene, it allows easy visual inspection while protecting contents from humidity. Desiccant and desiccator grease are available separately.
  • Stackable, transparent polystyrene construction.
  • Foam rubber door gasket and two roller latches for an airtight fit.
  • Inside dimensions: 21 x 18 x 13 cm (8.25 x 7 x 5 in).
  • Door opening: 19 x 11 cm (7.5 x 4.4 in).
  • Desiccant and desiccator grease sold separately.
  • Quantity: 1 each.
Catalog No. NC3781094 Supplier Hach Company Supplier No. 2618100
The desiccator cabinet, non-vacuum provides an airtight, stackable storage solution for moisture-sensitive samples and components. Constructed of transparent polystyrene, it allows easy visual inspection while protecting contents from humidity. Desiccant and desiccator grease are available separately.
  • Stackable, transparent polystyrene construction.
  • Foam rubber door gasket and two roller latches for an airtight fit.
  • Inside dimensions: 21 x 18 x 13 cm (8.25 x 7 x 5 in).
  • Door opening: 19 x 11 cm (7.5 x 4.4 in).
  • Desiccant and desiccator grease sold separately.
  • Quantity: 1 each.