Learn More
MSC DESTACO Flatfoot Vacuum Cup, 3 inch, Nitrile, 45 Durometer, B-Mount
Catalog No. 5000532000
Encompass
These flatfoot vacuum cups are designed for use on moderately flat to medium contour, dry surfaces. They offer significant holding capacities both horizontally and vertically. Constructed from durable nitrile material with a 45 durometer, these vacuum cups ensure a strong grip and include a 3/8 inch NPT mount option.
  • Constructed from durable nitrile material with a 45 durometer.
  • Equipped with a 3/8 inch NPT mount.
  • Measures 76.2 mm (3.00 in) in diameter.
  • Lightweight at 57.0 g (2.0 oz).
  • Resistant to oil, water, grease, hydraulic fluid, and alcohol.
  • Operating temperature range from -29° to 121°C (-20° to 250°F) for short durations.
  • Horizontal dry holding capacity of 30.0 lbf.
  • Vertical lift holding capacity of 60 lbf.
Catalog No. 50-005-32000 Supplier MSC Supplier No. 70043104
These flatfoot vacuum cups are designed for use on moderately flat to medium contour, dry surfaces. They offer significant holding capacities both horizontally and vertically. Constructed from durable nitrile material with a 45 durometer, these vacuum cups ensure a strong grip and include a 3/8 inch NPT mount option.
  • Constructed from durable nitrile material with a 45 durometer.
  • Equipped with a 3/8 inch NPT mount.
  • Measures 76.2 mm (3.00 in) in diameter.
  • Lightweight at 57.0 g (2.0 oz).
  • Resistant to oil, water, grease, hydraulic fluid, and alcohol.
  • Operating temperature range from -29° to 121°C (-20° to 250°F) for short durations.
  • Horizontal dry holding capacity of 30.0 lbf.
  • Vertical lift holding capacity of 60 lbf.