Learn More
MSC DESTACO Round Bellows Vacuum Cup, Nitrile, 3.00 in Cup Size
Catalog No. 5000515189
Encompass
This round bellows vacuum cup is constructed from durable Nitrile, designed for effective handling applications. It features a 3.00 in cup size and provides strong holding capabilities in various environments.
  • Constructed from Nitrile for durability and chemical resistance.
  • Features a 3.00 in cup size for effective gripping.
  • Offers 12 lbf horizontal dry holding capacity.
  • Provides 6 lbf horizontal wet holding capacity.
  • Achieves 56 lbf vertical lift holding capacity.
  • Resistant to oil, water, grease, hydraulic fluid, and alcohol.
  • Suitable for various handling applications.
Catalog No. 50-005-15189 Supplier MSC Supplier No. 79635280
This round bellows vacuum cup is constructed from durable Nitrile, designed for effective handling applications. It features a 3.00 in cup size and provides strong holding capabilities in various environments.
  • Constructed from Nitrile for durability and chemical resistance.
  • Features a 3.00 in cup size for effective gripping.
  • Offers 12 lbf horizontal dry holding capacity.
  • Provides 6 lbf horizontal wet holding capacity.
  • Achieves 56 lbf vertical lift holding capacity.
  • Resistant to oil, water, grease, hydraulic fluid, and alcohol.
  • Suitable for various handling applications.