Learn More
Apexbio Technology LLC Desthiobiotin-6-dCTP 200ul (5 mM)
SDP
Catalog No. 5000091497
Encompass_Preferred
Desthiobiotin-6-dCTP is a modified deoxycytidine triphosphate derivative featuring a desthiobiotin moiety attached via a hexyl linker to the cytosine base It is designed to be efficiently incorporated into nucleic acids by DNA polymerases during enzymatic synthesis thereby enabling subsequent detection or isolation through streptavidin or anti-biotin antibody binding Based on these properties Desthiobiotin-6-dCTP holds research potential in nucleic acid probe labeling in situ hybridization assays genetic testing nucleic acid purification and other molecular biology research applications
Catalog No. 50-000-91497 Supplier Apexbio Technology LLC Supplier No. B81560.2
Desthiobiotin-6-dCTP is a modified deoxycytidine triphosphate derivative featuring a desthiobiotin moiety attached via a hexyl linker to the cytosine base It is designed to be efficiently incorporated into nucleic acids by DNA polymerases during enzymatic synthesis thereby enabling subsequent detection or isolation through streptavidin or anti-biotin antibody binding Based on these properties Desthiobiotin-6-dCTP holds research potential in nucleic acid probe labeling in situ hybridization assays genetic testing nucleic acid purification and other molecular biology research applications