Learn More
Medchemexpress LLC Desthiobiotin-Iodoacetamide | 2924824-04-2 | C14H25IN4O3 | 25 MG
SDP
Catalog No. 5000374245
Encompass_Preferred
Desthiobiotin-Iodoacetamide is a chemical compound that functions as an ADC linker, facilitating the attachment of cytotoxins to antibodies in antibody-drug conjugates. It also serves as a probe for labeling Oridonin-treated cell lysis. This product is for research use only and has not been fully validated for medical applications.
  • Functions as an ADC linker
  • Acts as a probe for labeling cell lysis treated with Oridonin
  • Purity of 99.51% (LCMS)
  • Appears as a white to off-white solid
Catalog No. 50-003-74245 Supplier Medchemexpress LLC Supplier No. 25MGHY150230
Desthiobiotin-Iodoacetamide is a chemical compound that functions as an ADC linker, facilitating the attachment of cytotoxins to antibodies in antibody-drug conjugates. It also serves as a probe for labeling Oridonin-treated cell lysis. This product is for research use only and has not been fully validated for medical applications.
  • Functions as an ADC linker
  • Acts as a probe for labeling cell lysis treated with Oridonin
  • Purity of 99.51% (LCMS)
  • Appears as a white to off-white solid