Learn More
Medchemexpress LLC Desthiobiotin-Iodoacetamide | 2924824-04-2 | C14H25IN4O3 | 50 MG
SDP
Catalog No. 5000374767
Encompass_Preferred
Desthiobiotin-Iodoacetamide serves as an ADC linker, a crucial component in antibody-drug conjugates. This compound can function as a probe to label cell lysis treated with Oridonin. It is supplied as a white to off-white solid with a high purity of 99.51%.
  • Functions as an ADC linker
  • Can be used as a probe for Oridonin-treated cell lysis
  • Relevant for research in cancer, targeted therapy, and immunotherapy
  • Solid appearance, white to off-white
  • Purity of 99.51%
Catalog No. 50-003-74767 Supplier Medchemexpress LLC Supplier No. HY15023050MG
Desthiobiotin-Iodoacetamide serves as an ADC linker, a crucial component in antibody-drug conjugates. This compound can function as a probe to label cell lysis treated with Oridonin. It is supplied as a white to off-white solid with a high purity of 99.51%.
  • Functions as an ADC linker
  • Can be used as a probe for Oridonin-treated cell lysis
  • Relevant for research in cancer, targeted therapy, and immunotherapy
  • Solid appearance, white to off-white
  • Purity of 99.51%