Learn More
Medchemexpress LLC Desthiobiotin-NHS ester | 80750-24-9 | MFCD08062086 | >98.0% | 311.33 | C14H21N3O5 | 1 G
SDP
Catalog No. 5000281657
Encompass_Preferred
Desthiobiotin-NHS ester is an NHS-activated desthiobiotin reagent that reacts with primary amines to form stable amide bonds, enabling reversible affinity capture with streptavidin in labeling and bioconjugation workflows. Supplied as a dry powder with recommended low-temperature storage, it is used for protein labeling, biochemical assays, and metalloprotein probing.
  • Reacts with primary amines to form stable amide bonds.
  • Enables reversible streptavidin-based capture due to lower affinity than biotin.
  • Suitable for protein labeling, bioconjugation, and biochemical assays.
  • Supplied as a powder with recommended low-temperature storage.
  • High purity for reliable conjugation performance.
Catalog No. 50-002-81657 Supplier Medchemexpress LLC Supplier No. HYW4377451G
Desthiobiotin-NHS ester is an NHS-activated desthiobiotin reagent that reacts with primary amines to form stable amide bonds, enabling reversible affinity capture with streptavidin in labeling and bioconjugation workflows. Supplied as a dry powder with recommended low-temperature storage, it is used for protein labeling, biochemical assays, and metalloprotein probing.
  • Reacts with primary amines to form stable amide bonds.
  • Enables reversible streptavidin-based capture due to lower affinity than biotin.
  • Suitable for protein labeling, bioconjugation, and biochemical assays.
  • Supplied as a powder with recommended low-temperature storage.
  • High purity for reliable conjugation performance.