Learn More
Medchemexpress LLC Desthiobiotin-NHS ester | 80750-24-9 | MFCD08062086 | 98.0% | 311.33 g/mol | C14H21N3O5 | 10 MG
SDP
Catalog No. 5000280536
Encompass_Preferred
Desthiobiotin-NHS ester is an N-hydroxysuccinimide (NHS)-activated biotin analog used to covalently label primary amines on proteins and other biomolecules. The reagent reacts with lysine residues to form stable amide bonds, enabling affinity capture and detection with streptavidin-based tools.
  • NHS-activated ester reacts selectively with primary amines.
  • Forms stable amide bonds for durable conjugation.
  • Enables streptavidin-based affinity capture and detection.
  • Suitable for protein labeling, pull-down assays, and probe development.
  • High purity, commonly supplied at 98.0%.
  • Available in small lab-scale quantities such as 10 mg.
Catalog No. 50-002-80536 Supplier Medchemexpress LLC Supplier No. HYW43774510MG
Desthiobiotin-NHS ester is an N-hydroxysuccinimide (NHS)-activated biotin analog used to covalently label primary amines on proteins and other biomolecules. The reagent reacts with lysine residues to form stable amide bonds, enabling affinity capture and detection with streptavidin-based tools.
  • NHS-activated ester reacts selectively with primary amines.
  • Forms stable amide bonds for durable conjugation.
  • Enables streptavidin-based affinity capture and detection.
  • Suitable for protein labeling, pull-down assays, and probe development.
  • High purity, commonly supplied at 98.0%.
  • Available in small lab-scale quantities such as 10 mg.