Learn More
Medchemexpress LLC Desthiobiotin-NHS ester | 80750-24-9 | MFCD08062086 | 98.0% | 311.33 g/mol | C14H21N3O5 | 250 MG
SDP
Catalog No. 5000281358
Encompass_Preferred
Desthiobiotin-NHS ester is an amine-reactive biotin analogue supplied as an N-hydroxysuccinimide (NHS) ester for covalent labeling of primary amines on proteins and other biomolecules. It facilitates reversible affinity capture and is intended for biochemical assays, probe preparation, and affinity purification workflows.
Catalog No. 50-002-81358 Supplier Medchemexpress LLC Supplier No. HYW437745250MG
Desthiobiotin-NHS ester is an amine-reactive biotin analogue supplied as an N-hydroxysuccinimide (NHS) ester for covalent labeling of primary amines on proteins and other biomolecules. It facilitates reversible affinity capture and is intended for biochemical assays, probe preparation, and affinity purification workflows.