Learn More
Medchemexpress LLC Desthiobiotin-PEG4-acid | 461.55 | 50 MG
SDP
Catalog No. 5000366201
Encompass_Preferred
Desthiobiotin-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-66201 Supplier Medchemexpress LLC Supplier No. 50MGHY138507
Desthiobiotin-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins