Learn More
Medchemexpress LLC Desthiobiotin-PEG4-acid | >98% | 461.55 | 25 MG
SDP
Catalog No. 5000348459
Encompass_Preferred
Desthiobiotin-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used as a PEG-based PROTAC linker
  • Useful in the synthesis of PROTACs
  • Enables selective degradation of target proteins
Catalog No. 50-003-48459 Supplier Medchemexpress LLC Supplier No. 25MGHY138507
Desthiobiotin-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used as a PEG-based PROTAC linker
  • Useful in the synthesis of PROTACs
  • Enables selective degradation of target proteins