Learn More
Medchemexpress LLC Desthiobiotin polyethyleneoxide iodoacetamide | 97.7% | C21H38IN5O6 | 1 MG
SDP
Catalog No. 5000354351
Encompass_Preferred
Desthiobiotin polyethyleneoxide iodoacetamide is a chemical compound primarily utilized for cysteine labeling in chemoproteomic experiments. It is intended for research use only.
  • Utilized for cysteine labeling in chemoproteomic experiments
  • Intended for research use only
Catalog No. 50-003-54351 Supplier Medchemexpress LLC Supplier No. HY1432051MG
Desthiobiotin polyethyleneoxide iodoacetamide is a chemical compound primarily utilized for cysteine labeling in chemoproteomic experiments. It is intended for research use only.
  • Utilized for cysteine labeling in chemoproteomic experiments
  • Intended for research use only