Learn More
Medchemexpress LLC Desthiobiotin polyethyleneoxide iodoacetamide | 97.7% | C21H38IN5O6 | 100 MG
SDP
Catalog No. 5000354718
Encompass_Preferred
Desthiobiotin polyethyleneoxide iodoacetamide is a compound useful for cysteine labeling in chemoproteomic experiments. It is for research use only and not sold to patients.
  • Useful for cysteine labeling in chemoproteomic experiments
  • Available in solid form
  • White to light yellow in color
  • Molecular weight of 583.46
  • Store powder at -20°C for 3 years
  • Unstable in solutions, freshly prepared solutions recommended
Catalog No. 50-003-54718 Supplier Medchemexpress LLC Supplier No. HY143205100MG
Desthiobiotin polyethyleneoxide iodoacetamide is a compound useful for cysteine labeling in chemoproteomic experiments. It is for research use only and not sold to patients.
  • Useful for cysteine labeling in chemoproteomic experiments
  • Available in solid form
  • White to light yellow in color
  • Molecular weight of 583.46
  • Store powder at -20°C for 3 years
  • Unstable in solutions, freshly prepared solutions recommended