Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Desthiobiotin polyethyleneoxide iodoacetamide | 97.7% | C21H38IN5O6 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY14320550MG

Encompass_Preferred

Desthiobiotin polyethyleneoxide iodoacetamide is a chemical compound utilized for cysteine labeling in chemoproteomic experiments. It is supplied as a white to light yellow solid, recommended for storage as a powder at -20°C for up to 3 years.

  • Utilized for cysteine labeling in chemoproteomic experiments.
  • Supplied as a solid, white to light yellow in appearance.
  • Recommended storage as a powder at -20°C for 3 years.
  • Unstable in solutions; freshly prepared solutions are recommended.
  • Soluble in DMSO at 100 mg/mL.

Catalog No. 50-003-54977


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.