Learn More
Medchemexpress LLC Desthiobiotin polyethyleneoxide iodoacetamide | MFCD06202009 | 97.7% | 583.46 | C21H38IN5O6 | 5 MG
SDP
Catalog No. 5000221763
Encompass_Preferred
Desthiobiotin polyethyleneoxide iodoacetamide is an iodoacetamide-based probe that labels cysteine residues by alkylation, introducing a desthiobiotin affinity tag through a polyethylene oxide linker for enrichment and downstream mass spectrometry-based chemoproteomic analysis.
  • Selectively alkylates cysteine thiols for targeted labeling.
  • Introduces a desthiobiotin affinity handle for streptavidin-based enrichment.
  • Polyethylene oxide linker improves solubility and reduces steric hindrance.
  • Compatible with chemoproteomic workflows and mass spectrometry analysis.
  • Stable as a powder when stored at -20°C for extended periods.
Catalog No. 50-002-21763 Supplier Medchemexpress LLC Supplier No. HY1432055MG
Desthiobiotin polyethyleneoxide iodoacetamide is an iodoacetamide-based probe that labels cysteine residues by alkylation, introducing a desthiobiotin affinity tag through a polyethylene oxide linker for enrichment and downstream mass spectrometry-based chemoproteomic analysis.
  • Selectively alkylates cysteine thiols for targeted labeling.
  • Introduces a desthiobiotin affinity handle for streptavidin-based enrichment.
  • Polyethylene oxide linker improves solubility and reduces steric hindrance.
  • Compatible with chemoproteomic workflows and mass spectrometry analysis.
  • Stable as a powder when stored at -20°C for extended periods.