Learn More
GA International Destructible Cryogenic Labels for Vials & Containers 1" x 0.5" / 25.4mm x 12.7mm Rectangle White From -196°C to +100°C (-321°F to +212°F)
Catalog No. 5000571435
Encompass_Preferred
Destructible labels, designed for the secure identification of cryo vials, microtubes, and other containers. This includes cardboard and cryo boxes, plates, vials, and where visible evidence of label tampering is required, such as in pharmaceutical applications. Once adhered to the surface, the labels cannot be removed without being destroyed and broken into small pieces. These cryogenic labels will withstand prolonged storage in laboratory freezers, transport on dry ice, and thawing in water baths. Can be printed with thermal-transfer printers or handwritten with our permanent cryogenic markers.
Catalog No. 50-005-71435 Supplier GA International Supplier No. CTDB7C11WH
Destructible labels, designed for the secure identification of cryo vials, microtubes, and other containers. This includes cardboard and cryo boxes, plates, vials, and where visible evidence of label tampering is required, such as in pharmaceutical applications. Once adhered to the surface, the labels cannot be removed without being destroyed and broken into small pieces. These cryogenic labels will withstand prolonged storage in laboratory freezers, transport on dry ice, and thawing in water baths. Can be printed with thermal-transfer printers or handwritten with our permanent cryogenic markers.