Learn More
Cayman Chemical DestruxIn B2 100ug
SDP
Catalog No. 502988043
Encompass_Preferred
A mycotoxin with antiviral, insecticidal, and phytotoxic activities; inhibits secretion of HBsAg by Hep3B cells expressing HBV DNA (IC50 = 1.3 µM); toxic to Sf9 insect cells (ECIS50 = 92 µM); phytotoxic to B. napus leaves
Catalog No. 50-298-8043 Supplier Cayman Chemical Supplier No. 30210100 UG
A mycotoxin with antiviral, insecticidal, and phytotoxic activities; inhibits secretion of HBsAg by Hep3B cells expressing HBV DNA (IC50 = 1.3 µM); toxic to Sf9 insect cells (ECIS50 = 92 µM); phytotoxic to B. napus leaves