Learn More
ATCC Streptococcus agalactiae
Supplier:  ATCC 51487
Encompass
Streptococcus agalactiae strain CIP 103768 was isolated from Tilapia sp. brain. This strain causes meningoencephalitis in fish.
Catalog No. 50-238-0467