Learn More
ATCC Desulfovibrio vulgaris; subsp. vulgaris
Catalog No. 502384050
Encompass
Desulfovibrio vulgaris subsp. vulgaris was isolated from Wealden clay in England. This whole-genome sequenced bacterial type strain can be used in biotechnology.
Catalog No. 50-238-4050 Supplier ATCC Supplier No. 29579
Desulfovibrio vulgaris subsp. vulgaris was isolated from Wealden clay in England. This whole-genome sequenced bacterial type strain can be used in biotechnology.