Learn More
Apexbio Technology LLC Desvenlafaxine Succinate 386750-22-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090811
Encompass_Preferred
Desvenlafaxine Succinate (CAS 386750-22-7) is a small molecule inhibitor that targets both the serotonin (5-HT) and norepinephrine (NE) transporters It acts by blocking the reuptake of these neurotransmitters thereby modulating synaptic concentrations In in vitro assays Desvenlafaxine Succinate demonstrates a high affinity for the serotonin transporter (K sub i /sub 40 2 nM) and a lower affinity for the norepinephrine transporter (K sub i /sub 558 4 nM) This compound is utilized in biomedical research to investigate the pharmacological mechanisms underlying monoaminergic neurotransmission and to study potential therapeutic strategies for neuropsychiatric conditions involving serotonergic and noradrenergic pathways
Catalog No. 50-000-90811 Supplier Apexbio Technology LLC Supplier No. B22465.1
Desvenlafaxine Succinate (CAS 386750-22-7) is a small molecule inhibitor that targets both the serotonin (5-HT) and norepinephrine (NE) transporters It acts by blocking the reuptake of these neurotransmitters thereby modulating synaptic concentrations In in vitro assays Desvenlafaxine Succinate demonstrates a high affinity for the serotonin transporter (K sub i /sub 40 2 nM) and a lower affinity for the norepinephrine transporter (K sub i /sub 558 4 nM) This compound is utilized in biomedical research to investigate the pharmacological mechanisms underlying monoaminergic neurotransmission and to study potential therapeutic strategies for neuropsychiatric conditions involving serotonergic and noradrenergic pathways