Learn More
TARGETMOL CHEMICALS INC Desvenlafaxine succinate hydra
SDP
Catalog No. 502880008
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Desvenlafaxine succinate hydrate (WY 45233 Succinate) is an antidepressant of the serotonin-norepinephrine reuptake inhibitor (SNRI). Purity 99.39%
Catalog No. 50-288-0008 Supplier TARGETMOL CHEMICALS INC Supplier No. T6188100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Desvenlafaxine succinate hydrate (WY 45233 Succinate) is an antidepressant of the serotonin-norepinephrine reuptake inhibitor (SNRI). Purity 99.39%