Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desyl bromide, 97%, Thermo Scientific™

Catalog No. AAH3269106
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3269106 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3269106 Supplier Thermo Scientific Chemicals Supplier No. H3269106

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1484-50-0
Molecular Formula C14H11BrO
Quantity 5g
MDL Number MFCD00000136
UN Number UN3261
Synonym 2-bromo-2-phenylacetophenone, desyl bromide, 2-bromo-1,2-diphenylethan-1-one, ethanone, 2-bromo-1,2-diphenyl, ethanone,2-bromo-1,2-diphenyl, 2-bromo-1,2-diphenyl-ethanone, desylbromide, alpha-bromodeoxybenzoin, alpha-bromodesoxibenzoine
InChI Key ZFFBIQMNKOJDJE-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(C(=O)C2=CC=CC=C2)Br
IUPAC Name 2-bromo-1,2-diphenylethanone
Molecular Weight (g/mol) 275.145
PubChem CID 102630
Formula Weight 275.14
Percent Purity 97%
Chemical Name or Material Desyl bromide
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 216-057-1
TSCA Yes
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.