Learn More
BIOCHEMICAL DIAGNOSTICS INC Stat-Skreen W/Mdma, Oxy & Bup, Positive 6X20Ml
Catalog No. NC1827557
Encompass_Preferred
External Liquid Ready to Use Control to validate the performance of URINE Drugs of Abuse Devices. 15 drug formulation in convenient 20ml fill. Positive control included in 6X20ml kitbox. 30 day open shelf life, stored 2-8C. Opiate cutoff level is 300ng/ml.
This item is not returnable. View return policy
Catalog No. NC1827557 Supplier BIOCHEMICAL DIAGNOSTICS INC Supplier No. 19466756 6PK
External Liquid Ready to Use Control to validate the performance of URINE Drugs of Abuse Devices. 15 drug formulation in convenient 20ml fill. Positive control included in 6X20ml kitbox. 30 day open shelf life, stored 2-8C. Opiate cutoff level is 300ng/ml.