Learn More
Roche Diagnostics DET SOL REFILL 4 X 60ML
Catalog No. 50589352 Shop All Roche Diagnostics Products
Encompass
Cedex Detergent is required for cleaning the capillary system of the Cedex and Cedex HiRes Systems. The ready-to-use solution is available as a component of the Cedex Reagent Kit and as a separate bottle.
Catalog No. 50-589-352 Supplier Roche Diagnostics Supplier No. 05650658001
Cedex Detergent is required for cleaning the capillary system of the Cedex and Cedex HiRes Systems. The ready-to-use solution is available as a component of the Cedex Reagent Kit and as a separate bottle.