Learn More
Cayman Chemical DETA NONOate, Purity: ≥95%, CAS: 146724-94-9, 100 mg
SDP
Catalog No. 502894514
Encompass_Preferred
DETA NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 20 hours and 56 hours at 37°C and 22-25°C, pH 7.4, respectively, to liberate 2 moles of NO per mole of parent compound.{1190,5437}
Catalog No. 50-289-4514 Supplier Cayman Chemical Supplier No. 82120100 MG
DETA NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 20 hours and 56 hours at 37°C and 22-25°C, pH 7.4, respectively, to liberate 2 moles of NO per mole of parent compound.{1190,5437}