Learn More
AdipoGen DETA NONOate,Chemical. CAS: 146724-94-9
SDP
Catalog No. 501146573
Encompass_Preferred
DETA NONOate, AdipoGen Life Sciences. Chemical. CAS: 146724-94-9. Formula: C4H13N5O2. MW: 163.2. Nitric oxide (NO) donor. The slow release of NO allows the use in experiments investigating the effects of prolonged constant NO delivery to cells and tissues. The half-life time is 20 hours at 37?C and 56 hours at 22-25?C in 0.1M phosphate buffer, pH 7.4. Liberates 2 mol of NO per mol of parent compound. Decomposition of NONOates is nearly instantaneous at pH 5.0.
Catalog No. 50-114-6573 Supplier AdipoGen Supplier No. AGCR13614M025
DETA NONOate, AdipoGen Life Sciences. Chemical. CAS: 146724-94-9. Formula: C4H13N5O2. MW: 163.2. Nitric oxide (NO) donor. The slow release of NO allows the use in experiments investigating the effects of prolonged constant NO delivery to cells and tissues. The half-life time is 20 hours at 37?C and 56 hours at 22-25?C in 0.1M phosphate buffer, pH 7.4. Liberates 2 mol of NO per mol of parent compound. Decomposition of NONOates is nearly instantaneous at pH 5.0.