Learn More
AdipoGen DETA NONOate (5 mg)
SDP
Supplier:  AdipoGen AGCR13614M005
Encompass_Preferred
Chemical. CAS: 146724-94-9. Formula: C4H13N5O2. MW: 163.2. Nitric oxide (NO) donor. The slow release of NO allows the use in experiments investigating the effects of prolonged constant NO delivery to cells and tissues. The half-life time is 20 hours at 37°C and 56 hours at 22-25°C in 0.1M phosphate buffer, pH 7.4. Liberates 2 mol of NO per mol of parent compound. Decomposition of NONOates is nearly instantaneous at pH 5.0.
Catalog No. 50-205-3982