Learn More
Cole-Parmer Cole-Parmer Detachable cable; BNC, 10 ft (Electrode Cable)
Catalog No. 50633597 Shop All Cole-Parmer Products
Encompass_Preferred
This 5-foot long NC cable is used to connect electrodes to meters, controllers and transmeters that accept BNC connections.
Catalog No. 50-633-597 Supplier Cole-Parmer Supplier No. 0566109
This 5-foot long NC cable is used to connect electrodes to meters, controllers and transmeters that accept BNC connections.