Learn More
Amsbio LLC Detachin Cell Detachment Solution
Supplier:  Amsbio LLC AMS.T100110
Encompass
Detachin™ Cell Detachment Solution is a superior alternative to Trypsin/EDTA for gently detaching adherent cells from in vitro growth vessels. Detachin™ provides quick, gentle, and effective detachment of a wide variety of adherent cells, including primary cells, from all known tissue culture plastic ware. It contains protease and collagenase activities in an isotonic, phosphate buffer solution with EDTA.
Catalog No. 50-204-1278