Learn More
Enzo Life Sciences Diethyldithiocarbamic acid. sodium salt. trihydrate (high purity) (25g). CAS: 20624-25-3
SDP
Supplier:  Enzo Life Sciences ALX400003G025
Encompass_Preferred
Nitric oxide (NO) spin-trapping reagent. Thiol and iron chelator. Inhibits induction of macrophage nitric oxide synthase (NOS). Has been shown to be an inhibitor of the nuclear transcription factor κB (NF-κB). Alternative name: DETC (high purity), Diethyldithiocarbamate (high purity). Purity: ≥99% (Titration). Solubility: Soluble in water (20mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9066