Learn More
BURKLE INC Detectable liquid funnel PS, Ø 3.94 in. (100 mm), blue, sterile
Catalog No. 503081271
Encompass_Preferred
The detectable disposable funnel, made of blue polystyrene, is produced in cleanroom class 7 (10,000), individually packaged for single use, and sterilized.
Besides its blue color, it contains an additive that makes it visible to metal detectors and X-rays, allowing rejection by standard foreign object detection systems despite being plastic.
Ideal for reliably filling bottles and containers with narrow openings, the funnel prevents liquid or free-flowing solid loss during transfer.
Produced in cleanrooms, it features a suspension eye and venting fins to ensure uniform flow while pouring.
Combined with filter paper, the funnel is also suitable for filtering applications.
Catalog No. 50-308-1271 Supplier BURKLE INC Supplier No. 53786004
The detectable disposable funnel, made of blue polystyrene, is produced in cleanroom class 7 (10,000), individually packaged for single use, and sterilized.
Besides its blue color, it contains an additive that makes it visible to metal detectors and X-rays, allowing rejection by standard foreign object detection systems despite being plastic.
Ideal for reliably filling bottles and containers with narrow openings, the funnel prevents liquid or free-flowing solid loss during transfer.
Produced in cleanrooms, it features a suspension eye and venting fins to ensure uniform flow while pouring.
Combined with filter paper, the funnel is also suitable for filtering applications.