Learn More
BURKLE INC Volumetric spoon 0.85 oz. (25 ml), PS blue detectable, sterile
Catalog No. 503081186
Encompass_Preferred
In addition to its blue color, these samplers contain a special additive making them visible to metal detectors and X-rays. Though plastic, they can be detected and removed by standard foreign body detection systems.
Measuring and dosing spoons allow precise measurement of powders, granules, and liquids. Excess is scraped off the top edge, with volume embossed on the handle.
The flat front edge is perfect for scooping from container bottoms and corners, while the flat base allows spill-free placement.
Designed for single use, these disposable spoons eliminate cleaning needs and prevent cross-contamination, making them ideal for sampling small quantities.
Catalog No. 50-308-1186 Supplier BURKLE INC Supplier No. 53784097
In addition to its blue color, these samplers contain a special additive making them visible to metal detectors and X-rays. Though plastic, they can be detected and removed by standard foreign body detection systems.
Measuring and dosing spoons allow precise measurement of powders, granules, and liquids. Excess is scraped off the top edge, with volume embossed on the handle.
The flat front edge is perfect for scooping from container bottoms and corners, while the flat base allows spill-free placement.
Designed for single use, these disposable spoons eliminate cleaning needs and prevent cross-contamination, making them ideal for sampling small quantities.