Learn More
BURKLE INC Detectable scoops, long handle, 3.38 oz. (100 ml), PS, sterile
Catalog No. 503081218
Encompass_Preferred
These samplers are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
The sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content or to reach through narrow openings.
With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container.
Catalog No. 50-308-1218 Supplier BURKLE INC Supplier No. 53785020
These samplers are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
The sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content or to reach through narrow openings.
With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container.