Learn More
BURKLE INC Detectable scoops, blue, PS, sterile, 5.07 oz. (150 ml)
Catalog No. 503081262
Encompass_Preferred
Detectable SteriPlast scoops, made of blue polystyrene, are produced in our class 7 clean room (10,000), individually packaged for disposable use and then sterilised by gamma rays.
Detectable scoops are ideal for powders, granulates and pastes.
These samplers are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
Catalog No. 50-308-1262 Supplier BURKLE INC Supplier No. 53782007
Detectable SteriPlast scoops, made of blue polystyrene, are produced in our class 7 clean room (10,000), individually packaged for disposable use and then sterilised by gamma rays.
Detectable scoops are ideal for powders, granulates and pastes.
These samplers are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.