Learn More
BURKLE INC Detectable spoons, blue, PS, sterile, 2,5 ml
Catalog No. 503081243
Encompass_Preferred
Detectable SteriPlast spoons, made of polystyrene, are produced in our class 7 clean room (10,000), individually packaged for disposable use and then sterilised by gamma rays.
The detectable spoons are available in two sizes, 0.08 oz. (2.5 ml) (similar to a teaspoon) and 10 ml (similar to a tablespoon), and are ideal for sampling powders, granulates and fluids.
These samplers are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
Catalog No. 50-308-1243 Supplier BURKLE INC Supplier No. 53782011
Detectable SteriPlast spoons, made of polystyrene, are produced in our class 7 clean room (10,000), individually packaged for disposable use and then sterilised by gamma rays.
The detectable spoons are available in two sizes, 0.08 oz. (2.5 ml) (similar to a teaspoon) and 10 ml (similar to a tablespoon), and are ideal for sampling powders, granulates and fluids.
These samplers are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.