Learn More
BURKLE INC Detectable spoons, long handle, 0.68 oz. (20ml), PS, sterile
Catalog No. 503081232
Encompass_Preferred
In addition to the blue color, these samplers have a special additive in the material that makes them visible in metal detectors or by X-ray. Although they are made of plastic, these products can therefore be sorted out using the usual systems for checking for foreign bodies.
The sample spoon with long handle can be used to remove powders, granulates, pastes or liquids.
The long handle also makes it easy to reach the bottoms of larger containers or to sample drums, canisters and bottles with small opening diameters. The notches on the handle make the disposable spoon comfortable and safe to hold.
Catalog No. 50-308-1232 Supplier BURKLE INC Supplier No. 53785008
In addition to the blue color, these samplers have a special additive in the material that makes them visible in metal detectors or by X-ray. Although they are made of plastic, these products can therefore be sorted out using the usual systems for checking for foreign bodies.
The sample spoon with long handle can be used to remove powders, granulates, pastes or liquids.
The long handle also makes it easy to reach the bottoms of larger containers or to sample drums, canisters and bottles with small opening diameters. The notches on the handle make the disposable spoon comfortable and safe to hold.