Learn More
BIOCHEMICAL DIAGNOSTICS INC Negative Liq Urine Control, 6X20Ml
Catalog No. NC1827558
Encompass_Preferred
External Liquid Ready to Use NEGATIVE Control to validate the performance of URINE Drugs of Abuse Devices. NEGATIVE control included in 6X20ml kitbox. 30 day open shelf life, stored 2-8C.
This item is not returnable. View return policy
Catalog No. NC1827558 Supplier BIOCHEMICAL DIAGNOSTICS INC Supplier No. 19223011 6PK
External Liquid Ready to Use NEGATIVE Control to validate the performance of URINE Drugs of Abuse Devices. NEGATIVE control included in 6X20ml kitbox. 30 day open shelf life, stored 2-8C.