Learn More
MOLECULAR DEPOT LLC Detection Buffer CLIA - 50 mL
Catalog No. 5000013418
Encompass_Preferred
Detection Antibody Stabilization Buffer consists of a chemiluminescence buffer extensively tested for the stabilization of a wide variety of detection conjugates (proteins and antibodies) This buffer contains a proprietary collection of proteins detergents and other chemicals that promote the long-term stability of magnetic beads For in vitro research use only
Catalog No. 50-000-13418 Supplier MOLECULAR DEPOT LLC Supplier No. B2010188
Detection Antibody Stabilization Buffer consists of a chemiluminescence buffer extensively tested for the stabilization of a wide variety of detection conjugates (proteins and antibodies) This buffer contains a proprietary collection of proteins detergents and other chemicals that promote the long-term stability of magnetic beads For in vitro research use only