Learn More
Biogenex Laboratories NEW & IMPROVED SUPER SENSITIVE POLYMER-HRP IHC DETECTION KIT : 500 test
SDP
Supplier:  Biogenex Laboratories QD420YIKEN
Encompass_Preferred
his is a novel detection system using a non-biotin polymeric technology that makes use of two major components: Super Enhancer and a Poly-HRP reagent. As the system is not based on the biotin-avidin system, problems associated with endogenous biotin are completely eliminated. The technology allows excellent cell penetration ability for intense nuclear, cytoplasmic and membrane antigen staining when compared with other polymer-HRPs.
Catalog No. 50-223-8112