Learn More
Cell Signaling Technology Detection System Kit, LumiGLO™, Chemiluminescent, Western Blotting, 50 mL
SDP
Catalog No. 50194168
Encompass
This chemiluminescent substrate is a luminol-based system designed for use with HRP detection assays utilizing peroxidase-labeled antibodies immobilized on membranes. In the presence of hydrogen peroxide, horseradish peroxidase (HRP) converts luminol to an excited intermediate dianion that emits light upon returning to its ground state. Light emission is maximal immediately after exposure and continues for up to 3 hours, allowing for capture on X-ray film.
  • Detection of sub-picogram amounts of protein is routine with good primary antisera.
  • Entire detection procedure requires less than 1 hour.
  • Constant light emission for several minutes allows for multiple exposures to optimize signal intensity.
  • Re-exposure at a future date is achieved by adding more reagent.
  • Exposure times are typically seconds to minutes.
Catalog No. 50-194-168 Supplier Cell Signaling Technology Supplier No. 7003S
This chemiluminescent substrate is a luminol-based system designed for use with HRP detection assays utilizing peroxidase-labeled antibodies immobilized on membranes. In the presence of hydrogen peroxide, horseradish peroxidase (HRP) converts luminol to an excited intermediate dianion that emits light upon returning to its ground state. Light emission is maximal immediately after exposure and continues for up to 3 hours, allowing for capture on X-ray film.
  • Detection of sub-picogram amounts of protein is routine with good primary antisera.
  • Entire detection procedure requires less than 1 hour.
  • Constant light emission for several minutes allows for multiple exposures to optimize signal intensity.
  • Re-exposure at a future date is achieved by adding more reagent.
  • Exposure times are typically seconds to minutes.