Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Detection of the Influenza Virus Experiment Kit
SDPSCI_ED

Catalog No. S29012 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S29012 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29012 Supplier Edvotek™ Supplier No. 122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Horizontal gel electrophoresis apparatus, D.C. power supply, automatic micro pipettes and tips, balance, microwave or hot plate, pipette pump, 250 ml flasks or beakers, hot gloves, safety goggles and gloves, small plastic trays or large weigh boats, DNA visualization system (white light), distilled or deionized water

In this experiment, students perform two common tests (RIDT, RT-PCR) used to diagnose the flu in clinical setting. Edvotek™ Detection of the Influenza Virus Experiment Kit is for 8 lab groups.

Complete in 90 minutes

Specifications

Grade 9 to Undergraduate
Includes Ready-to-Load QuickStrip™ DNA samples, Flu Detection Strips, Flu Antibody Solution, Simulated Patient Samples, UltraSpec-Agarose™, Electrophoresis Buffer, FlashBlue™ DNA Stain, InstaStain™ Blue Cards, Microtipped Transfer Pipettes, Microcentrifuge Tubes
Time Range Complete in 90 minutes
Class Size 8 Lab Groups
Product Type Detection of the Influenza Virus Experiment Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.